PAS (with and without Digestion) Control Slides
Catalog # 77907-956
Supplier: VWR International
New Product
Specifications
- Description:PAS (with and without digestion) control slides
Specifications
About this item
PAS (with and without Digestion) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes 1 stained slide
Tissue: known positive
Special stain or antibody: PAS stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.