Hepatocellular Carcinoma Control Slides
Catalog # 77907-920
Supplier: VWR International
New Product
Specifications
- Description:Hepatocellular Carcinoma control slides
Specifications
About this item
Hepatocelluar Carcinoma control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included
Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.