ContactUsLinkComponent
Cotton Tip Applicator Stick
Cotton Tip Applicator Stick
Catalog # 77907-984
Supplier:  StatLab
New Product
Cotton Tip Applicator Stick
Catalog # 77907-984
Supplier:  StatLab
Supplier Number:  CTA-806W
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Cotton tip applicator stick
  • Length:
    6"
  • Environmentally Preferable:
Specifications
About this item
For application of tissue marking dyes and other routine uses in paraffin and frozen sectioning.
  • Splinter-free applicator stick with cotton tip
  • 6" length
  • Non sterile