Herpes Type II IHC Control Slides
Catalog # 77907-918
Supplier: VWR International
New Product
Specifications
- Description:Herpes type II IHC control slides
Specifications
About this item
Herpes Type II IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included Tissue: known positive
Special stain or antibody: Herpes Simplex Virus II
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.