Cytology Slides, 1 Circle
Catalog # 77907-952
Supplier: VWR International
New Product
Specifications
- Description:Cytology slides, with 1 circle
Specifications
About this item
Single circle cytology slides are 75×25 mm adhesion slides with a printed circle for positioning.
- Packaged 10 gross per case
Each well is placed in the exact position where the centrifuge specimen is deposited.