Disposable Plastic Lab Aprons
Polyethylene lab aprons
Catalog # 77908-140
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
Disposable polyethylene lab aprons.
Packed in dispenser box
Garment dimensions: 27×36"
Bib-style aprons are equipped with ties at the back for a secure fit.
Specifications
- Description:Polyethylene lab aprons