Aorta Control Slide
Aorta control slides
Catalog # 77907-868
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
Aorta control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
- Stained slide is not included
- Tissue: Aorta
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Specifications
- Description:Aorta control slides