ContactUsLinkComponent
Fungus PAS Control Slides
Fungus PAS Control Slides
  77907-908
 :  StatLab
New Product
undefined
Fungus PAS Control Slides
  77907-908
 :  StatLab
 :  CS-FUNGUS/PAS/25
New Product
  • Description:
    Fungus PAS control slides
Fungus PAS control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
    Tissue: known positive
    Special stain or antibody: PAS Stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.