Tonsil IHC Control Slides
Tonsil IHC control slides
Catalog # 77907-978
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
Tonsil IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
- Stained slide is not included Tissue: Tonsil
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.
Specifications
- Description:Tonsil IHC control slides