Orders LinkContactUsLinkComponent
PathFlow Supa Mega Cassettes
New Product
Supplier:   StatLab
PathFlow Supa Mega Cassettes
Supplier:  StatLab
New Product
Certificates
PathFlow Supa Mega Cassettes
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Supa Mega Cassette is particularly useful for prostate, brain, breast tissue, and eye. As a result, this cassette has been widely adopted by clinical, research, and toxicology laboratories.
  • Better reagent flow and improved turnaround times
    Proven writing surface
    Large color range available
The Supa Mega Cassette design offers better sectioning quality, with the innovative hex design ensuring a greater adhesion of the paraffin block to the cassette. This eliminates the possibility of the embedded specimen being separated from the cassette when sectioning fibrous specimens.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • 77908-012
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    Red
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $420.63
    $420.63
    Box of 100
       $420.63
    Supplier Number: EAG-0110-02A
    Specifications:
    New Product
    New Product
  • 77908-000
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    White
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $619.95
    $619.95
    Box of 100
       $619.95
    Supplier Number: EAG-0102-02A
    Specifications:
    New Product
    New Product
  • 77908-002
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    Green
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $619.95
    $619.95
    Box of 100
       $619.95
    Supplier Number: EAG-0104-02A
    Specifications:
    New Product
    New Product
  • 77908-004
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    Grey
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $619.95
    $619.95
    Box of 100
       $619.95
    Supplier Number: EAG-0105-02A
    Specifications:
    New Product
    New Product
  • 77908-006
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    Blue
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $619.95
    $619.95
    Box of 100
       $619.95
    Supplier Number: EAG-0106-02A
    Specifications:
    New Product
    New Product
  • 77908-008
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    Pink
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $619.95
    $619.95
    Box of 100
       $619.95
    Supplier Number: EAG-0108-02A
    Specifications:
    New Product
    New Product
  • 77908-010
    PathFlow Supa Mega cassettes
    PathFlow Supa Mega cassettes
    Yellow
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $619.95
    $619.95
    Box of 100
       $619.95
    Supplier Number: EAG-0109-02A
    Specifications:
    New Product
    New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)