20-Place Chipboard Slide Folder
Catalog # 77908-128
Supplier: VWR International
New Product
Specifications
- Description:20-place chipboard slide folder
Specifications
About this item
Design permits easy access and removal of slides.
20 slide capacity chipboard construction