ContactUsLinkComponent
ClariTex Supa Mega Microscope Slides, TwinFrost
ClariTex Supa Mega Microscope Slides, TwinFrost
Catalog # 77908-078
Supplier:  StatLab
New Product
ClariTex Supa Mega Microscope Slides, TwinFrost
Catalog # 77908-078
Supplier:  StatLab
Supplier Number:  MAD-1400-02A
New Product
Specifications
  • Description:
    ClariTex Supa Mega microscope slides, Twinfrost
  • L×W:
    52×76 mm
Specifications
About this item
Pre-cleaned and ready to use slides are an ideal time saving option.
  • Twinfrost slide
    1.0 to 1.2 mm square edge ground Manufactured to ISO 8037/1
The ClariTex Supa Mega microscope slides are double the average width of a standard slide, allowing sections to be much bigger and saving pathologists time spent switching between slides and fatigue from mentally reconstructing an image from several smaller sections.