ContactUsLinkComponent
Alcian Blue Control Slides
Catalog # 77907-872
Supplier:  VWR International
New Product
undefined
Alcian Blue Control Slides
Catalog # 77907-872
Supplier:  VWR International
New Product
Specifications
  • Description:
    Alcian Blue control slides
Specifications
About this item
Alcian Blue control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
    Tissue: known positive
    Special Stain Antibody: Alcian Blue pH 1.0/2.5
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.