Anti-DTYMK Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-405
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:DTYMK
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P23919
- Antigen synonyms:CDC8|Deoxythymidylate kinase (thymidylate kinase)|deoxythymidylate kinase|dTMP kinase|Dtymk|FLJ44192|KTHY_HUMAN|MGC198617|OTTHUMP00000164604|OTTHUMP00000200145|OTTHUMP00000200146|PP3731|Thymidylate kinase|TMPK|TYMK
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Thymidylate kinase protein (1-212AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: DTYMK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat