Anti-CTNNB1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-139
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CTNNB1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P35222
- Antigen synonyms:b-catenin|Beta catenin|Beta-catenin|Cadherin associated protein|Catenin (cadherin associated protein)|beta 1|88kDa|Catenin beta 1|Catenin beta-1|CATNB|CHBCAT|CTNB1_HUMAN|CTNNB|DKFZp686D02253|FLJ25606|FLJ37923|OTTHUMP00000162082|OTTHUMP00000165222|OTTHUMP00000165223|OTTHUMP00000209288|OTTHUMP00000209289
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Catenin beta-1 protein (2-781AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CTNNB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human