Anti-ADAM23 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77850-734
New Product
- Antigen symbol:ADAM23
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75077
- Antigen synonyms:MDC3Disintegrin and metalloproteinase domain-containing protein 23|ADAM 23|Metalloproteinase-like|disintegrin-like|and cysteine-rich protein 3|MDC-3
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Disintegrin and metalloproteinase domain-containing protein 23 protein (498-647AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: ADAM23
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human