Aspergillus niger Catalase
Catalog # CA80601-048
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:9001-05-2
- Species:Aspergillus niger
- Size:100 kU
- Storage conditions:Desiccated (hygroscopic), OK to freeze
- Enzyme name:Catalase
- Enzyme synonyms:Catalase