Anti-CBL Rabbit Polyclonal Antibody
100 µl, Vial
Catalog # 77847-898
Supplier: AFG Bioscience
Supplier #: A73401-100UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: CBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:100 µl
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Host:Rabbit
- Shipping temperature:Shipped on blue ice
- Cat. No.:77847-898
- Reactivity:Human
- Gene ID:P22681
- Antigen synonyms:4732447J05Rik|C CBL|Cas Br M (murine) ecotropic retroviral transforming sequence|Casitas B lineage lymphoma proto oncogene|Casitas B-lineage lymphoma proto-oncogene|CBL 2|cbl|CBL_HUMAN|CBL2|E3 ubiquitin protein ligase CBL|E3 ubiquitin-protein ligase CBL|Oncogene CBL2|Proto oncogene c CBL|Proto-oncogene c-CBL|RGD1561386|RING finger protein 55|RNF55|Signal transduction protein CBL
- Immunogen:Peptide sequence around aa. 698~702 (T-E-Y-M-T) derived from Human c-Cbl.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:CBL
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- ELISA:Yes



