Anti-COQ6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-118
- 77846-119
- Antigen symbol:
- COQ6
- COQ6
- Antigen synonyms:
- CGI-10|Coenzyme Q6 homolog (yeast)|Coenzyme Q6 homolog|monooxygenase (S. cerevisiae)|monooxygenase (yeast)|coq6|COQ6_HUMAN|Ubiquinone biosynthesis monooxygenase COQ6
- CGI-10|Coenzyme Q6 homolog (yeast)|Coenzyme Q6 homolog|monooxygenase (S. cerevisiae)|monooxygenase (yeast)|coq6|COQ6_HUMAN|Ubiquinone biosynthesis monooxygenase COQ6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Z9
- Q9Y2Z9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquinone biosynthesis monooxygenase COQ6, mitochondrial protein (208-339AA)
- Recombinant Human Ubiquinone biosynthesis monooxygenase COQ6, mitochondrial protein (208-339AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together