Anti-CRNN Rabbit Recombinant Polyclonal Antibody
77853-584
New Product
- Antigen symbol:CRNN
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UBG3
- Antigen synonyms:53 kDa putative calcium binding protein|53 kDa putative calcium-binding protein|53 kDa squamous epithelial induced stress protein|53 kDa squamous epithelial-induced stress protein|58 kDa heat shock protein|C1orf10|Chromosome 1 open reading frame 10|Cornulin|CRNN_HUMAN|DRC1|PDRC1|SEP53|Squamous epithelial heat shock protein 53|Tumor related protein|Tumor-related protein
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cornulin protein (139-389AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: CRNN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat