Anti-ATP6V0A2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ATP6V0A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-375
- 77846-376
- Antigen symbol:
- ATP6V0A2
- ATP6V0A2
- Antigen synonyms:
- a2|A2V ATPase|ARCL|ATP6a2|ATP6N1D|ATPase, H+ transporting, lysosomal V0 subunit a isoform 2|ATPase, H+ transporting, lysosomal V0 subunit a2|Infantile malignant osteopetrosis|J6B7|Lysosomal H(+) transporting ATPase V0 subunit a2|Lysosomal H(+)-transporting ATPase V0 subunit a2|regeneration and tolerance factor|Stv1|TJ6|TJ6M|TJ6s|V ATPase 116 kDa isoform a2|V type proton ATPase 116 kDa subunit a|V type proton ATPase 116 kDa subunit a isoform 2|V-ATPase 116 kDa isoform a2|V-type proton ATPase 116 kDa subunit a isoform 2|Vacuolar proton translocating ATPase 116 kDa subunit a|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2|Vph1|VPP2_HUMAN|WSS
- a2|A2V ATPase|ARCL|ATP6a2|ATP6N1D|ATPase, H+ transporting, lysosomal V0 subunit a isoform 2|ATPase, H+ transporting, lysosomal V0 subunit a2|Infantile malignant osteopetrosis|J6B7|Lysosomal H(+) transporting ATPase V0 subunit a2|Lysosomal H(+)-transporting ATPase V0 subunit a2|regeneration and tolerance factor|Stv1|TJ6|TJ6M|TJ6s|V ATPase 116 kDa isoform a2|V type proton ATPase 116 kDa subunit a|V type proton ATPase 116 kDa subunit a isoform 2|V-ATPase 116 kDa isoform a2|V-type proton ATPase 116 kDa subunit a isoform 2|Vacuolar proton translocating ATPase 116 kDa subunit a|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2|Vph1|VPP2_HUMAN|WSS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 2 protein (476-549AA)
- Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 2 protein (476-549AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together