Anti-CHTOP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHTOP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-165
- 77846-166
- Antigen symbol:
- CHTOP
- CHTOP
- Antigen synonyms:
- C1orf77|chromatin target of chromatin target of PRMT1 protein|Chromatin target of PRMT1|Chromatin target of PRMT1 protein|Chromosome 1 open reading frame 77|CHTOP_HUMAN|DKFZp547E1010|FL- RP1-178F15.2|FL-SRAG|FOP|Friend of PRMT1|Friend of PRMT1 protein|HT031|MGC131924|MGC86949|OTTHUMP00000035122|OTTHUMP00000035123|Pp7704|RP1 178F15.2|Small arginine- and glycine-rich protein|Small protein rich in arginine and glycine|SRAG|SRAG-3|SRAG-5
- C1orf77|chromatin target of chromatin target of PRMT1 protein|Chromatin target of PRMT1|Chromatin target of PRMT1 protein|Chromosome 1 open reading frame 77|CHTOP_HUMAN|DKFZp547E1010|FL- RP1-178F15.2|FL-SRAG|FOP|Friend of PRMT1|Friend of PRMT1 protein|HT031|MGC131924|MGC86949|OTTHUMP00000035122|OTTHUMP00000035123|Pp7704|RP1 178F15.2|Small arginine- and glycine-rich protein|Small protein rich in arginine and glycine|SRAG|SRAG-3|SRAG-5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3Y2
- Q9Y3Y2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Chromatin target of PRMT1 protein (179-248AA)
- Recombinant human Chromatin target of PRMT1 protein (179-248AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together