Anti-ACTL7B Rabbit Recombinant Polyclonal Antibody
Catalog # 77846-629
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ACTL7B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen synonyms:ACL7B_HUMAN|Actin like 7 beta|Actin like 7B|Actin like protein 7B|Actin-like protein 7B|Actin-like-7-beta|MGC151192|MGC151194|OTTHUMP00000021867|OTTMUSP00000007742|RP11-3J11.2|RP23-48F4.1|t actin 1|Tact1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Actin-like protein 7B protein (1-415AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ACTL7B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse