Anti-SLC39A7 Rabbit Polyclonal Antibody
77847-115
New Product
- Antigen symbol:SLC39A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q92504
- Antigen synonyms:D6S115E|D6S2244E|H2 KE4|Histidine rich membrane protein Ke4|Histidine-rich membrane protein Ke4|HKE4|HLA class II region expressed gene KE4|KE4|Ke4 gene mouse, human homolog of|Really interesting new gene 5 protein|RING5|S39A7_HUMAN|SLC39A7|Solute carrier family 39 (zinc transporter), member 7|Solute carrier family 39 member 7|solute carrier family 39 zinc transporter member 7|Zinc transporter SLC39A7|ZIP7
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human SLC39A7.
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: SLC39A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse