Anti-ARL6IP5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-400
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ARL6IP5
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75915
- Antigen synonyms:Addicsin|ADP ribosylation factor like protein 6 interacting protein 5|ADP ribosylation like factor 6 interacting protein 5|ADP-ribosylation factor-like protein 6-interacting protein 5|Aip 5|Aip-5|Aip5|ARL 6 interacting protein 5|ARL-6-interacting protein 5|Cytoskeleton related vitamin A responsive protein|Cytoskeleton-related vitamin A-responsive protein|Dermal papilla derived protein 11|Dermal papilla-derived protein 11|DERP 11|DERP11|Glutamate transporter EAAC1-interacting protein|Glutamate transporter EEAC 1 associated protein|Glutamate transporter EEAC1 associated protein|GTRAP3 18|GTRAP3-18|Hp 22|Hp22|HSPC 127|HSPC127|JM 5|JM5|JMX|JWA|PRA 1 domain family 3|PRA 2|PRA1 domain family 3|PRA1 family protein 3|PRA2|PRAF 3|PRAF3|PRAF3_HUMAN|Prenylated Rab acceptor protein 2|Protein JWa|Putative MAPK activating protein PM27|Putative MAPK-activating protein PM27
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human PRA1 family protein 3 protein (136-188AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ARL6IP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human