Anti-NFE2L2 (phospho-Ser40) HEK293F cells Recombinant Monoclonal Antibody [clone: 2E6]
77850-373
New Product
- Antigen symbol:NFE2L2
- Clonality:Recombinant Monoclonal
- Clone:2E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q16236
- Antigen synonyms:Nuclear factor erythroid 2-related factor 2, NF-E2-related factor 2, NFE2-related factor 2, HEBP1, Nuclear factor, erythroid derived 2, like 2, NFE2L2, NRF2
- Modification:phospho-Ser40
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-NFE2L2 (S40)
- Purification:Affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:50 to 1:200, IF: 1:20 to 1:200
Type:
Antigen: NFE2L2
Clonality: Recombinant Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human