Anti-TP53BP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: TP53BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-450
- 77848-449
- Antigen symbol:
- TP53BP2
- TP53BP2
- Antigen synonyms:
- 53BP2|Apoptosis stimulating of p53 protein 2|Apoptosis stimulating protein of p53 2|Apoptosis-stimulating of p53 protein 2|ASPP2|ASPP2_HUMAN|BBP|Bcl2 binding protein|Bcl2-binding protein|NY REN 51 antigen|p53 binding protein 2|p53-binding protein 2|p53BP2|PPP1R13A|Renal carcinoma antigen NY-REN-51|Tumor protein p53 binding protein 2|Tumor suppressor p53 binding protein 2|Tumor suppressor p53-binding protein 2
- 53BP2|Apoptosis stimulating of p53 protein 2|Apoptosis stimulating protein of p53 2|Apoptosis-stimulating of p53 protein 2|ASPP2|ASPP2_HUMAN|BBP|Bcl2 binding protein|Bcl2-binding protein|NY REN 51 antigen|p53 binding protein 2|p53-binding protein 2|p53BP2|PPP1R13A|Renal carcinoma antigen NY-REN-51|Tumor protein p53 binding protein 2|Tumor suppressor p53 binding protein 2|Tumor suppressor p53-binding protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13625
- Q13625
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TP53BP2
- Fusion protein of Human TP53BP2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together