Anti-LOXL2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LOXL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-189
- 77846-190
- Antigen symbol:
- LOXL2
- LOXL2
- Antigen synonyms:
- LOR 2|LOR2|LOX L2|LOXL 2|LOXL2|LOXL2_HUMAN|Lysyl oxidase homolog 2|Lysyl oxidase like 2|Lysyl oxidase like protein 2|Lysyl oxidase related 2|Lysyl oxidase related protein 2|Lysyl oxidase related protein WS9 14|Lysyl oxidase-like protein 2|Lysyl oxidase-related protein 2|Lysyl oxidase-related protein WS9-14|WS9 14
- LOR 2|LOR2|LOX L2|LOXL 2|LOXL2|LOXL2_HUMAN|Lysyl oxidase homolog 2|Lysyl oxidase like 2|Lysyl oxidase like protein 2|Lysyl oxidase related 2|Lysyl oxidase related protein 2|Lysyl oxidase related protein WS9 14|Lysyl oxidase-like protein 2|Lysyl oxidase-related protein 2|Lysyl oxidase-related protein WS9-14|WS9 14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4K0
- Q9Y4K0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysyl oxidase homolog 2 protein (27-121AA)
- Recombinant Human Lysyl oxidase homolog 2 protein (27-121AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together