Anti-APOBEC2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:1000 to 1:2000, WB:1:200 to 1:1000
Type:
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77846-700
- 77846-701
- Antigen symbol:
- APOBEC2
- APOBEC2
- Antigen synonyms:
- APOBEC2C->U-editing enzyme APOBEC-2|EC 3.5.4.36|mRNA(cytosine(6666)) deaminase 2
- APOBEC2C->U-editing enzyme APOBEC-2|EC 3.5.4.36|mRNA(cytosine(6666)) deaminase 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y235
- Q9Y235
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human APOBEC2
- Synthetic peptide of Human APOBEC2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together