Anti-CCR3 Rabbit Recombinant Polyclonal Antibody
77852-556
New Product
- Antigen symbol:CCR3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:P51677
- Antigen synonyms:B chemokine receptor|b-chemokine receptor|C C chemokine receptor type 3|C C CKR 3|C-C chemokine receptor type 3|C-C CKR-3|CC chemokine receptor 3|CC chemokine receptor type 3|CC CKR 3|CC R3|CC-CKR-3|CCCKR3|CCR 3|CCR-3|CCR3_HUMAN|CD 193|CD193|CD193 antigen|Chemokine (C C motif) receptor 3|chemokine (C-C motif) receptor 3|Chemokine (CC) receptor 1 like 2|Chemokine CC motif receptor 3|Chemokine receptor 3|CK R3|CKR 3|CKR3|CMKB R3|CMKBR 3|CMKBR1L2|CMKBR3|Eosinophil CC chemokine receptor 3|Eosinophil eotaxin receptor|Eotaxin receptor|Macrophage inflammatory protein 1 alpha receptor like 2|MGC102841|MIP1 Alpha RL2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human C-C chemokine receptor type 3 protein (306-355AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CCR3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse