Anti-GTF2IRD1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GTF2IRD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-315
- 77843-314
- Antigen symbol:
- GTF2IRD1
- GTF2IRD1
- Antigen synonyms:
- CREAM1|General transcription factor II I repeat domain containing protein 1|General transcription factor II-I repeat domain-containing protein 1|General transcription factor III|GT2D1_HUMAN|GTF2I repeat domain containing protein 1|GTF2I repeat domain-containing protein 1|GTF2IRD1|GTF3|Muscle TFII I repeat domain-containing protein 1|Muscle TFII-I repeat domain-containing protein 1|MUSTRD1|MusTRD1/BEN|RBAP2|Slow muscle fiber enhancer binding protein|Slow-muscle-fiber enhancer-binding protein|USE B1 binding protein|USE B1-binding protein|WBSCR11|WBSCR12|Williams Beuren syndrome chromosome region 11 protein|Williams-Beuren syndrome chromosomal region 11 protein|Williams-Beuren syndrome chromosomal region 12 protein
- CREAM1|General transcription factor II I repeat domain containing protein 1|General transcription factor II-I repeat domain-containing protein 1|General transcription factor III|GT2D1_HUMAN|GTF2I repeat domain containing protein 1|GTF2I repeat domain-containing protein 1|GTF2IRD1|GTF3|Muscle TFII I repeat domain-containing protein 1|Muscle TFII-I repeat domain-containing protein 1|MUSTRD1|MusTRD1/BEN|RBAP2|Slow muscle fiber enhancer binding protein|Slow-muscle-fiber enhancer-binding protein|USE B1 binding protein|USE B1-binding protein|WBSCR11|WBSCR12|Williams Beuren syndrome chromosome region 11 protein|Williams-Beuren syndrome chromosomal region 11 protein|Williams-Beuren syndrome chromosomal region 12 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHL9
- Q9UHL9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human General transcription factor II-I repeat domain-containing protein 1 protein (436-560AA)
- Recombinant Human General transcription factor II-I repeat domain-containing protein 1 protein (436-560AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together