Anti-DOK5 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77854-321
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GRPEL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9HAV7
- Antigen synonyms:FLJ25609|GREPEL 1|GREPEL1|GrpE like 1|GrpE like 1 mitochondrial|GrpE like protein cochaperone|GrpE protein homolog 1 mitochondrial|GrpE protein homolog 1|mitochondrial|GRPE1_HUMAN|GRPEL 1|Grpel1|HMGE|Mt GrpE#1|Mt-GrpE#1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Mt-GrpE#1
- Purification:Antigen affinity purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human