Anti-ADRA2A Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-824
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ADRA2A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P08913
- Antigen synonyms:ADA2A_HUMAN|ADRA2|ADRA2R|ADRAR|Adrenergic alpha 2A receptor|Alpha 2 adrenergic receptor subtype C10|Alpha 2A adrenergic receptor|Alpha 2AAR subtype C10|Alpha-2 adrenergic receptor subtype C10|alpha-2-adrenergic receptor|platelet type|Alpha-2A adrenergic receptor|Alpha-2A adrenoceptor|Alpha-2A adrenoreceptor|Alpha-2AAR|ALPHA2AAR|OTTHUMP00000058849|ZNF32
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alpha-2A adrenergic receptor protein (218-374AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: ADRA2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human