Anti-LPAR2 Rabbit Polyclonal Antibody
Catalog # 77859-085
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:LPAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9HBW0
- Antigen synonyms:EDG 4|EDG-4|EDG4|Endothelial Cell Differentiation Gene 4|Endothelial differentiation lysophosphatidic acid G protein coupled receptor 4|FLJ93869|G protein coupled receptor|IPA2|LPA 2|LPA receptor 2|LPA receptor EDG4|LPA-2|LPA2|LPAR2|LPAR2_HUMAN|Lysophosphatidic acid receptor 2|Lysophosphatidic acid receptor Edg 4|Lysophosphatidic acid receptor Edg-4|Lysophosphatidic acid receptor EDG4
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence from Human Lysophosphatidic acid receptor 2 protein (8-27AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:500 to 1:1000
Type:
Antigen: LPAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human