Anti-ERCC6L2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-187
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ERCC6L2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:Q5T890
- Antigen synonyms:C9orf102|Chromosome 9 open reading frame 102|excision repair cross-complementing rodent repair deficiency|complementation group 6-like 2|FLJ37706|MGC30192|MGC43364|Putative DNA repair and recombination protein RAD26-like|putative repair and recombination helicase RAD26L|RAD26_HUMAN|Rad26l|RAD26L hypothetical protein|SR278|stretch responsive protein 278
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA excision repair protein ERCC-6-like 2 protein (72-371AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ERCC6L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human