Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Caspase Inhibitor VI
Caspase Inhibitor VI
Caspase Inhibitor VI
Catalog # CA80108-892
Supplier:
MilliporeSigma
CAS Number:
Certificates
Expand image
Caspase Inhibitor VI
Catalog # CA80108-892
Supplier:
MilliporeSigma
Supplier Number:
219007-1MG
CAS Number:
Certificates
Specifications
Pack type:
Plastic ampoule
CAS No.:
161401-82-7
Size:
1 mg
Storage conditions:
Safe to freeze
Enzyme name:
Caspase Inhibitor VI
Purity:
≥98% (HPLC)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation