Rat Fibronectin (from Plasma)
Catalog # CA80051-670
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:86088-83-7
- Source:Plasma
- Species:Rat
- Size:500 µg
- Storage conditions:Safe to freeze, avoid repeated freeze/thaw cycles
- Protein/peptide name:Fibronectin