Anti-CRADD Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-523
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CRADD
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P78560
- Antigen synonyms:CASP2 and RIPK1 domain containing adaptor with death domain|Caspase and RIP adapter with death domain|Caspase and RIP adaptor with death domain|CRADD_HUMAN|Death adaptor molecule RAIDD|Death domain containing protein Death domain-containing protein MGC9163|RIP associated ICH1/CED3 homologous protein with death domain|RIP associated protein with a death domain|RIP-associated protein with a death domain
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Death domain-containing protein CRADD protein (1-199AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: CRADD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human