Anti-SIPA1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: SIPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77848-466
- 77848-467
- Antigen symbol:
- SIPA1
- SIPA1
- Antigen synonyms:
- GTPase activating protein Spa 1|GTPase-activating protein Spa-1|MGC102688|MGC17037|p130 SPA-1|p130 SPA1|Signal induced proliferation associated 1|Signal induced proliferation associated gene 1|Signal induced proliferation associated protein 1|Signal-induced proliferation-associated protein 1|SIPA 1|Sipa-1|Sipa1|SIPA1_HUMAN|Spa1
- GTPase activating protein Spa 1|GTPase-activating protein Spa-1|MGC102688|MGC17037|p130 SPA-1|p130 SPA1|Signal induced proliferation associated 1|Signal induced proliferation associated gene 1|Signal induced proliferation associated protein 1|Signal-induced proliferation-associated protein 1|SIPA 1|Sipa-1|Sipa1|SIPA1_HUMAN|Spa1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FS4
- Q96FS4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SIPA1
- Synthetic peptide of Human SIPA1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together