Anti-ADD3 Rabbit Polyclonal Antibody
77848-963
New Product
- Antigen symbol:ADD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UEY8
- Antigen synonyms:ADDG_HUMAN|ADDL|Adducin 3 (gamma)|Adducin 3|Adducin|gamma|Adducin-like|Adducin-like protein 70|Gamma-adducin
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human ADD3.
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ADD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat