Anti-ATP6V1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-640
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ATP6V1A
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P38606
- Antigen synonyms:70kDa subunit|ATP6A1|ATP6V1A1|ATPase H+ transporting lysosomal subunit A1|ATPase H+ transporting|lysosomal 70kDa V1 subunit A|H(+) transporting two sector ATPase subunit A|H+ transporting ATPase chain A vacuolar (VA68 type)|HO68|OTTHUMP00000214746|V ATPase 69 kDa subunit 1|V ATPase A subunit 1|V ATPase subunit A 1|V-ATPase 69 kDa subunit|V-ATPase subunit A|V-type proton ATPase catalytic subunit A|VA68|Vacuolar ATP synthase catalytic subunit A ubiquitous isoform|Vacuolar ATPase isoform VA68|Vacuolar proton pump alpha subunit 1|Vacuolar proton pump subunit alpha|VATA_HUMAN|Vma1|VPP2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human V-type proton ATPase catalytic subunit A protein (159-405AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ATP6V1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human