Anti-ADAMTS5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: ADAMTS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-648
- 77847-649
- Antigen symbol:
- ADAMTS5
- ADAMTS5
- Antigen synonyms:
- A disintegrin and metalloproteinase with thrombospondin motifs 11|A disintegrin and metalloproteinase with thrombospondin motifs 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase 2)|A disintegrin-like and metalloprotease with thrombospondin type 1 motif, 5|A Disintigrin And Metalloproteinase with ThromboSpondin motif-5|ADAM metallopeptidase with thrombospondin type 1 motif 5|ADAM TS 11|ADAM TS 5|ADAM TS5|ADAM-TS 11|ADAM-TS 5|ADAM-TS5|ADAMTS 11|ADAMTS 5|ADAMTS-11|ADAMTS-5|ADAMTS11|ADAMTS11, formerly|Adamts5|ADMP 2|ADMP-2|ADMP2|Aggrecanase 2|Aggrecanase-2|ATS5_HUMAN|FLJ36738|Implantin|ThromboSpondin motif-5
- A disintegrin and metalloproteinase with thrombospondin motifs 11|A disintegrin and metalloproteinase with thrombospondin motifs 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase 2)|A disintegrin-like and metalloprotease with thrombospondin type 1 motif, 5|A Disintigrin And Metalloproteinase with ThromboSpondin motif-5|ADAM metallopeptidase with thrombospondin type 1 motif 5|ADAM TS 11|ADAM TS 5|ADAM TS5|ADAM-TS 11|ADAM-TS 5|ADAM-TS5|ADAMTS 11|ADAMTS 5|ADAMTS-11|ADAMTS-5|ADAMTS11|ADAMTS11, formerly|Adamts5|ADMP 2|ADMP-2|ADMP2|Aggrecanase 2|Aggrecanase-2|ATS5_HUMAN|FLJ36738|Implantin|ThromboSpondin motif-5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ADAMTS5
- Synthetic peptide of Human ADAMTS5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together