Anti-CBX5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-454
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CBX5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P45973
- Antigen synonyms:Antigen p25|CBX5|CBX5_HUMAN|CG8409|Chromobox 5|Chromobox homolog 5 (HP1 alpha homolog|Drosophila)|Chromobox homolog 5|Chromobox protein homolog 5|Epididymis luminal protein 25|HEL25|Heterochromatin protein 1 alpha|Heterochromatin protein 1|Heterochromatin protein 1 homolog alpha|HP1 alpha|HP1 alpha homolog|HP1|HP1A|HP1Hs alpha|Su(var)205
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Chromobox protein homolog 5 protein (1-191AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: CBX5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human