Anti-VIPAS39 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VIPAS39
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-233
- 77843-234
- Antigen symbol:
- VIPAS39
- VIPAS39
- Antigen synonyms:
- Apical basolateral polarity regulator|FLJ12707|hSPE-39|Protein spe-39 homolog|SPE 39|SPE 39 protein|SPE39|Uncharacterized protein C14orf133|vipar|VIPAR_HUMAN|VPS16B|VPS33B interacting protein|VPS33B interacting protein apical basolateral polarity regulator|VPS33B interacting protein involved in polarity and apical protein restriction|VPS33B-interacting protein
- Apical basolateral polarity regulator|FLJ12707|hSPE-39|Protein spe-39 homolog|SPE 39|SPE 39 protein|SPE39|Uncharacterized protein C14orf133|vipar|VIPAR_HUMAN|VPS16B|VPS33B interacting protein|VPS33B interacting protein apical basolateral polarity regulator|VPS33B interacting protein involved in polarity and apical protein restriction|VPS33B-interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H9C1
- Q9H9C1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Spermatogenesis-defective protein 39 homolog protein (1-493AA)
- Recombinant human Spermatogenesis-defective protein 39 homolog protein (1-493AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together