Anti-ERC1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-878
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ERC1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q8IUD2
- Antigen synonyms:Cast 2|CAST2|CAZ associated structural protein 2|Elks|ELKS/RAB6 interacting/CAST family member 1|ELKS/RAB6-interacting/CAST family member 1|ERC 1|ERC protein 1|ERC-1|ERC1 protein|KIAA1081|MGC12974|RAB6 interacting protein 2|RAB6-interacting protein 2|RAB6IP2|RB6I2_HUMAN
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human ELKS/Rab6-interacting/CAST family member 1 protein (1-224AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: ERC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human