Anti-PLA2G2D Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: PLA2G2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-338
- 77844-337
- Antigen symbol:
- PLA2G2D
- PLA2G2D
- Antigen synonyms:
- 2IID|EC 3.1.1.4|GIID sPLA2|Group IID secretory phospholipase A2|Group IID secretory phospholipase A2 precursor|PA2GD_HUMAN|Phosphatidylcholine 2 acylhydrolase GIID|Phosphatidylcholine 2-acylhydrolase 2D|Phospholipase A2 group IID|PLA2IID|Secretory phospholipase A2s|Secretory type PLA stroma associated homolog|Secretory-type PLA|sPLA|sPLA2-IID|sPLA2S|SPLASH|stroma-associated homolog
- 2IID|EC 3.1.1.4|GIID sPLA2|Group IID secretory phospholipase A2|Group IID secretory phospholipase A2 precursor|PA2GD_HUMAN|Phosphatidylcholine 2 acylhydrolase GIID|Phosphatidylcholine 2-acylhydrolase 2D|Phospholipase A2 group IID|PLA2IID|Secretory phospholipase A2s|Secretory type PLA stroma associated homolog|Secretory-type PLA|sPLA|sPLA2-IID|sPLA2S|SPLASH|stroma-associated homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNK4
- Q9UNK4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Group IID secretory phospholipase A2 protein (22-145AA)
- Recombinant Human Group IID secretory phospholipase A2 protein (22-145AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together