Endopeptidase Lys-C (from Achromobacter lyticus)
Catalog # CA60801-172
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- CAS No.:78642-25-8
- Source:Achromobacter lyticus
- Size:2 U
- Storage conditions:Safe to freeze, avoid repeated freeze/thaw cycles
- Enzyme name:Endopeptidase Lys-C