Anti-CTSC Rabbit Recombinant polyclonal Antibody
Catalog # 77853-752
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CTSC
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P53634
- Antigen synonyms:AI047818|CATC|CATC_HUMAN|Cathepsin C|Cathepsin J|CPPI|Dipeptidyl peptidase 1|Dipeptidyl peptidase 1 light chain|Dipeptidyl peptidase I|Dipeptidyl peptidase I exclusion domain chain|Dipeptidyl peptidase I heavy chain|Dipeptidyl peptidase I light chain|Dipeptidyl transferase|DPP I|DPP-I|DPPI|EC 3.4.14.1|HMS|JP|JPD|MGC126959 |PALS|PDON1|PLS
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Dipeptidyl peptidase 1 protein (25-134AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:200-1:500
Type:
Antigen: CTSC
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human