Anti-DUS4L Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-415
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:DUS4L
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O95620
- Antigen synonyms:DUS4LtRNA-dihydrouridine(20a/20b) synthase [NAD(P)+]-like|EC 1.3.1.-|pp35|tRNA-dihydrouridine synthase 4-like
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human tRNA-dihydrouridine(20a/20b) synthase [NAD(P)+]-like protein (1-196AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DUS4L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human