Anti-CLIC1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-248
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CLIC1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O00299
- Antigen synonyms:Chloride channel ABP|Chloride intracellular channel 1|Chloride intracellular channel protein 1|CLIC 1|CLIC1_HUMAN|G6|hRNCC|NCC 27|NCC27|Nuclear chloride ion channel 27|Nuclear chloride ion channel protein|p64CLCP|Regulatory nuclear chloride ion channel protein|RNCC protein
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Chloride intracellular channel protein 1 protein (42-241AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: CLIC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human